missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR44 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 25264-1-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
CRTH2 (chemoattractant receptor-homologous molecule), which is also known as the putative G protein-coupled receptor (GPR44), is expressed on Th2 cells and is a novel member of the G protein-coupled leukocyte chemoattractant receptor family, which is selectively expressed in Th2 but not Th1 lineage cells. CRTH2 is selectively expressed in vivo in an activated state of Th2 cells including allergen-responsive Th2 cells, suggesting its pivotal roles in ongoing Th2-type immune reactions.Especificaciones
GPR44 | |
Polyclonal | |
Unconjugated | |
PTGDR2 | |
CD294, CRTH2, DL1R, DP2, G protein coupled receptor 44, GPR44, Prostaglandin D2 Receptor 2, PTGDR2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11251 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9Y5Y4 | |
PTGDR2 | |
Peptide | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |