missing translation for 'onlineSavingsMsg'
Learn More

GNB3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

145.00€ - 410.00€

Especificaciones

Antígeno GNB3
Concentración 0.45 mg/mL
Aplicaciones Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Western Blot
Clasificación Polyclonal
Conjugado Unconjugated
Ver más especificaciones

Productos 2
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
16854544
Ver documentos
Proteintech
15388-1-AP-20UL
20 μL
145.00€
20 microlitros
Ver productos alternativos
Ahorre hasta 
»
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
16844544
Ver documentos
Proteintech
15388-1-AP-150UL
150 μL
410.00€
150 microlitros
Ver productos alternativos
Ahorre hasta 
»
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
Descripción

Descripción

Heterotrimeric guanine nucleotide-binding proteins in this gene is associated with essential hypertension and obesity. This polymorphism is also associated with the occurrence of the splice variant GNB3-s, which appears to have increased activity. GNB3-s is an example of alternative splicing caused by a nucleotide change outside of the splice donor and acceptor sites. Additional splice variants may exist for this gene, but they have not been fully described.
Especificaciones

Especificaciones

GNB3
Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Western Blot
Unconjugated
Rabbit
Mouse, Rat, Human
P16520, Q61011
14695, 2784, 60449
GNB3 Fusion Protein Ag7050
Primary
-20°C
GNB3
0.45 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
GNB3, Transducin beta chain 3
Gnb3
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documentos

Documentos

Product Certifications
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.