missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GNB3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 15388-1-AP-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Heterotrimeric guanine nucleotide-binding proteins in this gene is associated with essential hypertension and obesity. This polymorphism is also associated with the occurrence of the splice variant GNB3-s, which appears to have increased activity. GNB3-s is an example of alternative splicing caused by a nucleotide change outside of the splice donor and acceptor sites. Additional splice variants may exist for this gene, but they have not been fully described.Especificaciones
| GNB3 | |
| Polyclonal | |
| Unconjugated | |
| GNB3 | |
| GNB3, Transducin beta chain 3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 14695, 2784, 60449 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Western Blot | |
| 0.45 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P16520, Q61011 | |
| Gnb3 | |
| GNB3 Fusion Protein Ag7050 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido