Learn More
Glutamine synthetase Mouse anti-Human, Porcine, Clone: 1D10G8, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66323-1-IG-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Structural maintece of chromosomes (SMC) and non-SMC condensin proteins associate into complexes that have been implicated in the process of chromosome condensation. A crucial prerequisite for accurate segregation of replicated sister chromatids is the condensation of the chromosomes into a manageable form prior to metaphase. The condensin I complex consists of two SMC subunits, SMC2 and SMC4, and three non-SMC subunits, CAP-H, CAP-G, and CAP-D2. An alternative complex, the condensin II complex, contains alternate non-SMC subunits, CAP-G2, CAP-H2, and CAP-D3. CAP-H2 is also known as Non-SMC condensin II complex, subunit H2 (NCAPH2) or kleisin beta isoform 2. The three non-SMC subunits in the condensing complexes form a regulatory subcomplex that is required to activate the SMC ATPases and to promote mitosis-specific chromatin binding of the holocomplex. The precise individual functions of each non-SMC protein in activation remain to be determined.Especificaciones
Glutamine synthetase | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P15104, P46410 | |
GLUL | |
Glutamine synthetase Fusion Protein Ag6309 | |
150 μL | |
Primary | |
Human, Porcine | |
Antibody | |
IgG2b |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1D10G8 | |
Unconjugated | |
GLUL | |
GLNS, GLUL, Glutamate ammonia ligase, Glutamate decarboxylase, Glutamine synthetase, GS, PIG43, PIG59 | |
Mouse | |
Protein A | |
RUO | |
2752, 396944 | |
-20°C | |
Liquid |