Learn More
Varilla de carbono vítreo, 7 mm (0,28 in) de diámetro, tipo 1, Thermo Scientific Chemicals
Se emplea como material de electrodo en electroquímica. También se usa como crisol de alta temperatura y como componente de algunos dispositivos protésicos. Se utiliza como membrana selectiva de iones conductora de la electricidad.
Marca: Thermo Scientific Alfa Aesar 038012.DM
Detalles adicionales : CAS : 7440-44-0 Peso : 0.01000kg
Descripción
Glassy carbon rod is used as an electrode material in electrochemistry. It is also used as high temperature crucibles. Used as a component of some prosthetic devices. Used as electrical conductor ion-selective membrane.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Glassy carbon rod | |
MFCD00133992 | |
Insoluble in water. |
Tubo | |
100 mm |
RUO – Research Use Only