Learn More
Varilla de carbono vítreo, 6 mm (0,24 in) de diámetro, tipo 2, Thermo Scientific Chemicals
Marca: Thermo Scientific Alfa Aesar 045005.DM
Detalles adicionales : Peso : 0.01000kg
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Glassy carbon rod | |
Tubo | |
MFCD00133992 | |
100 mm |
RUO – Research Use Only