missing translation for 'onlineSavingsMsg'
Learn More

Varilla de carbono vítreo, 3 mm (0,1 in) de diámetro, tipo 1, Thermo Scientific Chemicals

Catalog Number p-7076437
Click to view available options
Cantidad:
50 mm
100 mm
200 mm
Este artículo no se puede devolver. Vea la política de devoluciones
Glassy carbon rod
7440-44-0
Tubo
MFCD00133992
Insoluble in water.
Este artículo no se puede devolver. Vea la política de devoluciones
Glassy carbon rod
7440-44-0
Tubo
MFCD00133992
Insoluble in water.

Se emplea como material de electrodo en electroquímica. También se emplea como material de electrodo para la fabricación de sensores. Sirve como material ideal en la tecnología de vacío. También se usa en técnicas de fundición y metalurgia, y en aleaciones duras y crisoles de alta temperatura.

Glassy carbon rod is used as an electrode material in electrochemistry. It is also employed as an electrode material for the fabrication of sensors. It serves as an ideal material for the vacuum technology. It is also used in casting, smelting of metal, hard alloys and high temperature crucibles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Fórmula molecular C
Cantidad 50 mm
Clave InChI OKTJSMMVPCPJKN-UHFFFAOYSA-N
SMILES [C]
Nombre IUPAC carbon
Peso molecular (g/mol) 12.01

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'