missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GEMIN7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 10625-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
The protein encoded by this gene is a component of the core SMN complex, which is required for pre-mRNA splicing in the nucleus. The encoded protein is found in the nucleoplasm, in nuclear "gems" (Gemini of Cajal bodies), and in the cytoplasm. Three transcript variants encoding the same protein have been found for this gene.Especificaciones
GEMIN7 | |
Polyclonal | |
Unconjugated | |
GEMIN7 | |
Gem associated protein 7, Gemin 7, GEMIN7, SIP3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
499391, 69731, 79760 | |
-20°C | |
Liquid |
ELISA | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9CWY4, Q9H840 | |
Gemin7 | |
GEMIN7 Fusion Protein Ag0976 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |