Learn More
GDA Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA553977
Detalles adicionales : Peso : 0.02000kg
Descripción
Immunogen sequence: AKTRDLHIQS HISENRDEVE AVKNLYPSYK NYTSVYDKNN LLTNKTVMAH GCYLSAEELN VFHERGASIA HCPNSNLSLS SGFLNVLEVL KHEVKIGLGT DVAG Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 88%.
GDA is an enzyme that catalyzes the hydrolytic deamination of guanine, producing xanthine and ammonia.Especificaciones
GDA | |
Polyclonal | |
Unconjugated | |
GDA | |
AU015411; AW047581; Cypin; cytoplasmic PSD95 interactor; GAH; Gda; GUANASE; guanine aminase; guanine aminohydrolase; Guanine deaminase; KIAA1258; NEDASIN; p51-nedasin | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9615 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y2T3 | |
GDA | |
Recombinant protein corresponding to Human GDA | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only