missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gamma cystathionase Mouse anti-Human, Mouse, Rat, Clone: 2C7F9, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 60234-1-IG-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a cytoplasmic enzyme in the trans-sulfuration pathway that converts cystathione derived from methionine into cysteine. Glutathione synthesis in the liver is dependent upon the availability of cysteine. Mutations in this gene cause cystathioninuria. Alternative splicing of this gene results in two transcript variants encoding different isoforms.Especificaciones
Gamma cystathionase | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P18757, P32929, Q8VCN5 | |
Cth | |
Gamma cystathionase Fusion Protein Ag2872 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
2C7F9 | |
Unconjugated | |
CTH | |
CSE, CTH, Cystathionine gamma lyase, Gamma cystathionase | |
Mouse | |
Protein G | |
RUO | |
107869, 1491, 24962 | |
-20°C | |
Liquid |