missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GALM Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 16022-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes an enzyme that catalyzes the epimerization of hexose sugars such as glucose and galactose. The encoded protein is expressed in the cytoplasm and has a preference for galactose. The encoded protein may be required for normal galactose metabolism by maintaining the equilibrium of alpha and beta anomers of galactose.Especificaciones
GALM | |
Polyclonal | |
Unconjugated | |
GALM | |
Aldose 1 epimerase, BLOCK25, Galactose mutarotase, GALM, IBD1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
130589, 313843, 319625 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q66HG4, Q8K157, Q96C23 | |
Galm | |
GALM Fusion Protein Ag8842 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |