Learn More
Ansell Edmont™ Marigold™ G07B Nitrile Gloves
Catalog Number 12959994
Click to view available options
Tamaño:
Small
Medium
Large
X-Large
Offers a food contact-approved nitrile formulation combined with a lightweight design. Ansell Edmont™ Marigold™ G07B Nitrile Gloves are an extremely strong yet sensitive glove.
  • 21CFR compliant, approved for all food contact
  • Raised diamond grip pattern for safe handling of food products
  • Tough and durable in use, with the comfort of latex
  • Lightweight construction for greater dexterity and ease in use
  • Recommended for use in food processing and preparation and janitorial work
Certificaciones/conformidad EN 388
EN 374 PT 2
EN 374 PT 3
21CFR
90/128/EEC
Color Blue
Material Nitrile
Palm Thickness (Metric) 0.22 mm
Tamaño Medium
Modelo 9834