missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FUT10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 18660-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Probable fucosyltransferase.Especificaciones
FUT10 | |
Polyclonal | |
Unconjugated | |
FUT10 | |
Fuc TX, Fucosyltransferase 10, Fucosyltransferase X, FucT X, FUT10 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
171167, 497619, 84750 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.24 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5F2L1, Q5F2L2, Q6P4F1 | |
Fut10 | |
FUT10 Fusion Protein Ag13415 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |