missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FTSJ1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11620-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
The protein encoded by this gene is a member of the S-adenosylmethionine-binding protein family. It is a nucleolar protein and may be involved in the processing and modification of rRNA. Three alternatively spliced transcript variants encoding different isoforms have been described for this gene.Especificaciones
FTSJ1 | |
Polyclonal | |
Unconjugated | |
FTSJ1 | |
CDLIV, FtsJ homolog 1 (E. coli), FTSJ1, JM23, MRX44, MRX9, Protein ftsJ homolog 1, SPB1, TRM7 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
24140, 363450, 54632 | |
-20°C | |
Liquid |
Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8BWQ4, Q9UET6 | |
Ftsj1 | |
FTSJ1 Fusion Protein Ag2199 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |