Learn More
Fisherbrand™ 3/8 in. Dot Labels
Fisher branded Cryogenic Storage Labels use a special adhesive that is designed to withstand extreme cold temperatures as low as -196°C (-311°F). Our cryo labels can be printed/written on and will not bleed, smudge or smear–perfect for identifying specimens after long periods of cryogenic storage.
Marca: Fisherbrand™
14.06 EUR hasta el 2024-05-31
Use el código promocional "21724" para aplicar la oferta.
Detalles adicionales : Peso : 0.18140kg
Descripción
- Labels can be printed/written on and will not bleed, smudge or smear - perfect for identifying specimens after long periods of cryogenic storage
Especificaciones
Rosa | |
No estéril | |
0,37 in |
Rollo de etiquetas de puntos | |
1000/rollo | |
9,55 mm |