missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher BioReagents™ Modified Terrific Broth (Capsules)
Used as a microbial growth medium for the cultivation of E. coli strains for the preparation of plasmid DNA and recombinant proteins.
114.93€
Especificaciones
Formato | Capsule |
---|---|
Tipo de medios | Terrific Broth |
Tipo de producto | Microbiology Media |
Formulación | Casein Peptone 12g/L, Yeast extract 24g/L, Potassium Phosphate, Monobasic 2.2g/L, Potassium Phosphate, Dibasic 9.4g/L |
Para utilizar con (aplicación) | For maintenance and propagation of Escherichia coli in molecular biology procedures |
Código de producto | Marca | Tipo de empaquetado | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Tipo de empaquetado | Precio | Cantidad y disponibilidad | |||||
11316002
|
Fisher BioReagents™
BP9729-600 |
Poly bottle, 25 Capsules (Each capsule contains 24g of broth) |
Este artículo actualmente no está disponible o ha sido discontinuado. Ver la página del producto para posibles alternativas |
||||||
Descripción
- Directions for Media Preparation: Add 2 capsules per 1L of purified water, such as BP2820 (Fisher BioReagents Microbial Cell Culture Grade Water). Then, add 4mL of glycerol to the mixture. Heat and frequently agitate to completely dissolve. Before use, sterilize by autoclaving for 15 minutes.
- Greater peptone and yeast concentration allow for higher plasmid yield
- Product is in capsule form
- Each capsule contains 24g of broth
- Format: Capsule
Especificaciones
Capsule | |
Microbiology Media | |
For maintenance and propagation of Escherichia coli in molecular biology procedures |
Terrific Broth | |
Casein Peptone 12g/L, Yeast extract 24g/L, Potassium Phosphate, Monobasic 2.2g/L, Potassium Phosphate, Dibasic 9.4g/L |