missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FBXO28 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 24282-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Members of the F-box protein family, such as FBXO28, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1.Especificaciones
FBXO28 | |
Polyclonal | |
Unconjugated | |
FBXO28 | |
F box only protein 28, F box protein 28, Fbx28, FBXO28, KIAA0483 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23219 | |
-20°C | |
Liquid |
Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9NVF7 | |
FBXO28 | |
FBXO28 Fusion Protein Ag14900 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |