Learn More
Invitrogen™ EVOS™ Onstage Incubator (OSI-2) Filter Replacement Kit
The EVOS Onstage Incubator (OSI-2) Filter Replacement Kit contains everything needed for the self-installation of new OSI-2 foam filters.
Marca:  Invitrogen™ AMEP4975
Detalles adicionales : Peso : 0.05000kg
Código de producto. 17856448
  • 67.20€ / cada uno
Fecha estimada de envío: 20-05-2024
para ver el stock
Descripción
Descripción
The EVOS Onstage Incubator (OSI-2) Filter Replacement Kit contains everything needed for the self-installation of new OSI-2 foam filters. These filters are recommended to be replaced every two years.
Especificaciones
Especificaciones
Onstage Accessory
EVOS™ OSI-2 Foam Filter Replacements
Two foam filters
EVOS
EVOS™ Onstage Incubator (OSI-2)
Room Temperature
Certificados
Ofertas especiales
Ofertas especiales
For Research Use Only. Not for use in diagnostic procedures.