Learn More
Invitrogen™ EVOS™ Onstage Incubator (OSI-2) Filter Replacement Kit
The EVOS Onstage Incubator (OSI-2) Filter Replacement Kit contains everything needed for the self-installation of new OSI-2 foam filters.
Marca: Invitrogen™ AMEP4975
Detalles adicionales : Peso : 0.05000kg
Descripción
The EVOS Onstage Incubator (OSI-2) Filter Replacement Kit contains everything needed for the self-installation of new OSI-2 foam filters. These filters are recommended to be replaced every two years.
Especificaciones
Onstage Accessory | |
EVOS™ OSI-2 Foam Filter Replacements | |
Two foam filters |
EVOS | |
EVOS™ Onstage Incubator (OSI-2) | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.