Learn More
Vileda Professional™ EvoControl™ 400 Microfiber Cleanroom Wipes
Código de producto. 17852476
Change view
Click to view available options
Dimensiones (L x An):
22,86 x 22,86 cm (9 x 9 in)
30,48 x 30,48 cm (12 x 12 in)
Tamaño de la unidad:
paquete de 1500
Código de producto. Dimensiones (L x An) unitSize
17852476 22,86 x 22,86 cm (9 x 9 in) paquete de 1500
17842476 30,48 x 30,48 cm (12 x 12 in) paquete de 1500
2 options
Código de producto. 17852476
Marca: Vileda Professional™ 142856
Solo quedan null
Añadir a la cesta
Añadir a la cesta
Designed for the removal of acids and chemical spills. Vileda Professional™ EvoControl™ 400 Microfiber Cleanroom Wipes are widely used in pharmaceutical, biotech and microelectronics industries.
  • Disposable liquid cleaning wipe
  • Clean glass and stainless steel without leaving residues or streaks
  • Microfiber material reach into microscopic surface pores to remove contamination thoroughly
  • Maximize particle pick up
  • Bacteria removal up to 99.999%
  • Cut fibers are melted together for sealed edge which prevents particle release
  • Disposable pre-laundered and laser cut 
  • Sealed edge for areas where moderate contamination control is required
Especificaciones
Tipo de producto Cleanroom Wipe
Material Microfibra
Color Blanco
Desechables Un solo uso
Bordes Laser cut
Dimensiones (L x An) 22,86 x 22,86 cm (9 x 9 in)
Cantidad de embalaje 150/Pk., 3000/Cs.