missing translation for 'onlineSavingsMsg'
Learn More

Etil (etoximetilen)cianoacetato, 98 %, Thermo Scientific Chemicals

Catalog Number p-7082095
Click to view available options
Cantidad:
50 g
250 g
1000 g
Este artículo no se puede devolver. Vea la política de devoluciones
94-05-3
C8H11NO3
169.18
MFCD00009136
KTMGNAIGXYODKQ-SREVYHEPSA-N
ethyl ethoxymethylene cyanoacetate, e-ethyl 2-cyano-3-ethoxyacrylate, ethyl 2-cyano-3-ethoxyacrylate, ethyl 3-ethoxy-2-cyanoacrylate, ethyl 2e-2-cyano-3-ethoxyprop-2-enoate, ethyl cyano ethoxymethylene acetate, 2-propenoic acid, 2-cyano-3-ethoxy-, ethyl ester, 2-cyano-3-ethoxy-2-propenoic acid ethyl ester, 2-cyano-3-ethoxyacrylic acid ethyl ester, ethyl 2e-2-cyano-3-ethoxy-2-propenoate
1715183
etil (E)-2-ciano-3-etoxiprop-2-enoato
CCO\C=C(\C#N)C(=O)OCC
Este artículo no se puede devolver. Vea la política de devoluciones
94-05-3
C8H11NO3
169.18
MFCD00009136
KTMGNAIGXYODKQ-SREVYHEPSA-N
ethyl ethoxymethylene cyanoacetate, e-ethyl 2-cyano-3-ethoxyacrylate, ethyl 2-cyano-3-ethoxyacrylate, ethyl 3-ethoxy-2-cyanoacrylate, ethyl 2e-2-cyano-3-ethoxyprop-2-enoate, ethyl cyano ethoxymethylene acetate, 2-propenoic acid, 2-cyano-3-ethoxy-, ethyl ester, 2-cyano-3-ethoxy-2-propenoic acid ethyl ester, 2-cyano-3-ethoxyacrylic acid ethyl ester, ethyl 2e-2-cyano-3-ethoxy-2-propenoate
1715183
etil (E)-2-ciano-3-etoxiprop-2-enoato
CCO\C=C(\C#N)C(=O)OCC

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Ethyl (ethoxymethylene)cyanoacetate
Punto de fusión 47°C to 52°C
Punto de ebullición 190°C to 191°C (30 mmHg)
Temperatura de inflamación 130°C (266°F)
Cantidad 50 g
Número UN UN3439
Beilstein 878814
Formula Weight (peso de la fórmula) 169.18
Porcentaje de pureza 98%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'