missing translation for 'onlineSavingsMsg'
Learn More

5-Cloroimidazo[1,2-a]piridina-2-carboxilato de etilo, 95 %, Thermo Scientific™

Catalog Number 15449015
Click to view available options
Cantidad:
250 mg
1 g
Este artículo no se puede devolver. Vea la política de devoluciones
67625-36-9
C10H9ClN2O2
224.644
MFCD09864797
UDQYTPCGXBXZCE-UHFFFAOYSA-N
ethyl 5-chloroimidazo 1,2-a pyridine-2-carboxylate, 5-chloro-imidazo 1,2-a pyridine-2-carboxylic acid ethyl ester, 2-ethoxycarbonyl-5-chloroimidazo 1,2-a pyridine, ethyl 5-chloroh-imidazo 1,2-a pyridine-2-carboxylate, ethyl 5-chloro-4-hydroimidazo 1,2-a pyridine-2-carboxylate, 5-chloroimidazo 1,2-a pyridine-2-carboxylic acid ethyl ester, imidazo 1,2-a pyridine-2-carboxylic acid,5-chloro-,ethyl ester
12428959
5-cloroimidazo[1,2-a]piridina-2-carboxilato de etilo
CCOC(=O)C1=CN2C(=N1)C=CC=C2Cl
Este artículo no se puede devolver. Vea la política de devoluciones
67625-36-9
C10H9ClN2O2
224.644
MFCD09864797
UDQYTPCGXBXZCE-UHFFFAOYSA-N
ethyl 5-chloroimidazo 1,2-a pyridine-2-carboxylate, 5-chloro-imidazo 1,2-a pyridine-2-carboxylic acid ethyl ester, 2-ethoxycarbonyl-5-chloroimidazo 1,2-a pyridine, ethyl 5-chloroh-imidazo 1,2-a pyridine-2-carboxylate, ethyl 5-chloro-4-hydroimidazo 1,2-a pyridine-2-carboxylate, 5-chloroimidazo 1,2-a pyridine-2-carboxylic acid ethyl ester, imidazo 1,2-a pyridine-2-carboxylic acid,5-chloro-,ethyl ester
12428959
5-cloroimidazo[1,2-a]piridina-2-carboxilato de etilo
CCOC(=O)C1=CN2C(=N1)C=CC=C2Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Ethyl 5-chloroimidazo[1,2-a]pyridine-2-carboxylate
Punto de fusión 79°C to 82°C
Cantidad 250 mg
Formula Weight (peso de la fórmula) 224.65
Porcentaje de pureza 95%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'