missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimetiltiazol-5-carboxilato de etilo, 97 %, Thermo Scientific™

Catalog Number 11489876
Click to view available options
Cantidad:
2.5 g
10 g
Este artículo no se puede devolver. Vea la política de devoluciones
7210-77-7
C8H11NO2S
185.24
MFCD00052874
BXOIIRQIGYJTTB-UHFFFAOYSA-N
ethyl 2,4-dimethylthiazole-5-carboxylate, 5-thiazolecarboxylic acid, 2,4-dimethyl-, ethyl ester, ethyl2,4-dimethylthiazole-5-carboxylate, pubchem15762, acmc-1bm0j, 5-ethoxycarbonyl-2,4-dimethylthiazole, ethyl-2,4-dimethylthiazole-5-carboxylate, 2,4-dimethyl-5-thiazolecarboxylic acid ethyl ester
603810
ethyl 2,4-dimethyl-1,3-thiazole-5-carboxylate
CCOC(=O)C1=C(C)N=C(C)S1
Este artículo no se puede devolver. Vea la política de devoluciones
7210-77-7
C8H11NO2S
185.24
MFCD00052874
BXOIIRQIGYJTTB-UHFFFAOYSA-N
ethyl 2,4-dimethylthiazole-5-carboxylate, 5-thiazolecarboxylic acid, 2,4-dimethyl-, ethyl ester, ethyl2,4-dimethylthiazole-5-carboxylate, pubchem15762, acmc-1bm0j, 5-ethoxycarbonyl-2,4-dimethylthiazole, ethyl-2,4-dimethylthiazole-5-carboxylate, 2,4-dimethyl-5-thiazolecarboxylic acid ethyl ester
603810
ethyl 2,4-dimethyl-1,3-thiazole-5-carboxylate
CCOC(=O)C1=C(C)N=C(C)S1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Ethyl 2,4-dimethylthiazole-5-carboxylate
Punto de fusión 48°C to 50°C
Cantidad 2.5 g
Formula Weight (peso de la fórmula) 185.24
Porcentaje de pureza 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'