missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EMILIN1 Mouse anti-Human, Clone: 2A5C8, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 60047-1-IG-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a protein that is one of the two components of elastic fibers. The encoded protein is rich in hydrophobic amino acids such as glycine and proline, which form mobile hydrophobic regions bounded by crosslinks between lysine residues. Deletions and mutations in this gene are associated with supravalvular aortic stenosis (SVAS) and autosomal domit cutis laxa. Multiple transcript variants encoding different isoforms have been found for this gene.Especificaciones
EMILIN1 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9Y6C2 | |
EMILIN1 | |
EMILIN1 Fusion Protein Ag1016 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
2A5C8 | |
Unconjugated | |
EMILIN1 | |
EMI, EMILIN, EMILIN 1, EMILIN1, gp115 | |
Mouse | |
Caprylic Acid Precipitation | |
RUO | |
11117 | |
-20°C | |
Liquid |