Learn More
ELAVL4 Mouse, anti-Human, Unconjugated, Clone: 1B1F5, Proteintech
Mouse Monoclonal Antibody
Marca:  Proteintech 67835-1-IG-150UL
Detalles adicionales : Peso : 0.00020kg
 Ver más versiones de este producto
Código de producto. 17201496
  • 325.38€ / 150 microlitros
Fecha estimada de envío: 04-10-2022
para ver el stock
Descripción
Descripción
ELAVL4, also named as HuD, is a 380 amino acid protein. The full name is neuronal embryonic lethal altered visual system-like(ELAVL) RNA-binding protein. ELAVL4 is expressed in brain and localized in the nucleus. PC12 cells expressing T7±HuD were treated with thio-specific (BMH) or amine-specific (DSS) chemical crosslinking reagents, and the resultant crosslinked complexes in the cell extracts were analyzed by immunoblotting with anti-T7 antibody. In addition to the monomeric HuD of 41 kDa, three specific crosslinked complexes could be detected using either of the crosslinkers. Two of the complexes had molecular masses of ∼80 and 120 kDa, and were likely to be the dimer and trimer of HuD, respectively. (PMID: 12384599 )
Especificaciones
Especificaciones
ELAVL4
Monoclonal
500 μg/mL
Mouse
Primary
Pig, Rabbit, Mouse, Rat, Human
ELISA, Immunohistochemistry, Western Blot
1B1F5
Unconjugated
150 μL
1996