missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EFCAB5 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 24296-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
The function of this protein remains unknown.Especificaciones
EFCAB5 | |
Polyclonal | |
Unconjugated | |
EFCAB5 | |
EFCAB5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
374786 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.55 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
A4FU69 | |
EFCAB5 | |
EFCAB5 Fusion Protein Ag19274 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |