missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ECHS1 Mouse anti-Human, Clone: 2B9D5, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66117-1-IG-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
The protein encoded by this gene functions in the second step of the mitochondrial fatty acid beta-oxidation pathway. It catalyzes the hydration of 2-trans-enoyl-coenzyme A (CoA) intermediates to L-3-hydroxyacyl-CoAs. The gene product is a member of the hydratase/isomerase superfamily. It localizes to the mitochondrial matrix. Transcript variants utilizing alternative transcription initiation sites have been described in the literature.Especificaciones
ECHS1 | |
Monoclonal | |
1.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P30084 | |
ECHS1 | |
ECHS1 Fusion Protein Ag16775 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Immunocytochemistry, Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence | |
2B9D5 | |
Unconjugated | |
ECHS1 | |
ECHS1, Enoyl CoA hydratase 1, SCEH | |
Mouse | |
Protein A | |
RUO | |
1892 | |
-20°C | |
Liquid |