missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ECEL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 14222-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a member of the M13 family of endopeptidases. In general, M13 family members are zinc-containing type II integral-membrane proteins that are important regulators of neuropeptide and peptide hormone activity. This gene is expressed specifically in the central nervous system and its protein localizes predominately to the endoplasmic reticulum or, in trace amounts, to the cell surface. Disruption of this gene in mouse embryonic stem cells results in neonatal lethality due to respiratory failure shortly after birth. Based on the specific expression of this gene and the phenotype of the gene deficiency in mouse embryos, it is suggested that this protein plays a critical role in neural regulation of the respiratory system. This gene has multiple pseudogenes.Especificaciones
ECEL1 | |
Polyclonal | |
Unconjugated | |
ECEL1 | |
DINE, ECEL1, ECEX, XCE, Xce protein | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
13599, 60417, 9427 | |
-20°C | |
Liquid |
ELISA | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O95672, Q9JHL3, Q9JMI0 | |
Ecel1 | |
ECEL1 Fusion Protein Ag5460 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |