Learn More
Thermo Scientific Pipetas electrónicas monocanal E1-ClipTip™, 1250
Electronic tip ejection, index finger pipetting action and ClipTip technology are ideal for repetitive pipetting tasks with microcentrifuge or test tubes.
Marca: Thermo Scientific 4670040
Detalles adicionales : Peso : 2.00000kg Transport : UN number : 3481 Chem class : 9
Descripción
Electronic tip ejection, index finger pipetting action and Thermo Scientitic™ ClipTip™ technology make the Thermo Scientific™ E1-ClipTip™ Electronic Single Channel Pipettes ideal for repetitive pipetting tasks with microcentrifuge or test tubes. Feel the difference.
Innovative ClipTip interlocking tip interface technology†- Tip is locked and sealed in place until ejected — no dropped tips that waste valuable samples
- Complete seal to secure the sample volume in each tip for enhanced accuracy and precision
- Tips are attached and ejected with a light touch — eliminates the frustration of banging tips on the pipette
- Reduce repetition and save time between labware formats with adjustable tip spacing in Equalizer models.
- Personalized user interface and password protection ensure consistent operation among multiple users.
- Designed for optimum comfort and ease of use, the electronic tip ejection and index finger pipetting action let your thumb relax during pipetting.
- Choose a step based Matrix or Presets with settings for your most common applications
- Customize your main menu view by organizing icons in your preferred order or creating shortcuts for your favorite programs and pipetting functions. You can also protect important settings from unintentional editing with a password.
- Tired of continuous editing? The Programs function allows you to store 20 your most common Matrix or Presets protocols with a specific name.
- Choose from single channels, 8- and 16-channel multichannels, or 6- and 12-channel adjustable tip spacing Equalizer multichannel pipettes within the volume range of 0.5-1250µL
- Service and calibration trackers remind you when the pipette needs to be sent to serviced
- Higher quality results with liquid specific calibrations
- Long lasting performance and fast recharging.
- Use either charging stand or a plug, you decide
- With convenient web registration
†E1-ClipTip 12.5 and all 384 E1-ClipTip pipettes utilize snap and lip tip fitting technology