Learn More
Nabertherm™ Drying oven
With their maximum working temperature of up to 300 °C and air circulation, the ovens achieve a perfect temperature uniformity which is much better than in ovens of most competitors. They can be used for various applications such as e.g. drying, sterilizing or warm storing.
Marca: Nabertherm™ TR-122BN
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
- TMAX: 300 °C
Especificaciones
B410 | |
300 °C | |
2 | |
900 × 610 × 860 mm |
120 l | |
7 | |
650 × 390 × 500 mm |