missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNM3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 14737-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Western blot using this antibody specifically detected the 100 kDa DNM3 in mouse brain. It doesn't cross-react with other DNMs.
The dynamins are a family of 100 kDa GTPases transcribed from at least three separate genes. At least four mRNA splice variants for each dynamin have been described. Dynamins contain several conserved regions including the conserved, amino-terminal GTPase domain, a centrally located membrane-binding plekstrin homology domain (PHD), and a coiled-coil region located in front of a proline-rich domain (PRD). The PRD is thought to mediate interactions between dynamin and numerous other cellular proteins. Dynamin I is expressed exclusively in neurons, dynamin II is ubiquitously expressed, and dynamin III is thought to be restricted to expression in the brain, testis, heart, and lung. The dynamins participate in the cellular process of clathrin-mediated and fluid-phase endocytosis.Especificaciones
DNM3 | |
Polyclonal | |
Unconjugated | |
DNM3 | |
DNM3, dynamin 3, Dynamin, testicular, KIAA0820, T dynamin | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
103967, 171574, 26052 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q08877, Q8BZ98, Q9UQ16 | |
Dnm3 | |
DNM3 Fusion Protein Ag6381 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |