missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAJC11 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 17331-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Belongs to the DNAJC11 family. Three named isoforms produced by alternative splicing.Especificaciones
DNAJC11 | |
Polyclonal | |
Unconjugated | |
DNAJC11 | |
DNAJC11 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
230935, 362666, 55735 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.14 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q5U458, Q9NVH1 | |
Dnajc11 | |
DNAJC11 Fusion Protein Ag11310 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |