missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ácido difenilditiofosfónico, Thermo Scientific Chemicals
Click to view available options
Cantidad:
0.5 g
2 g
10 g
Descripción
Diphenyldithiophosphonic acid is used in place of phenyl phosphate for acylation of nucleotides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Nombre del producto químico o material | Diphenyldithiophosphonic acid |
Punto de fusión | 55°C to 56°C |
Cantidad | 0.5 g |
Formula Weight (peso de la fórmula) | 250.31 |
Forma física | cristalino/a/s |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'