missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl ether, For pesticide residue analysis, Honeywell™ Riedel-de Haën™
Click to view available options
Cantidad:
1 L
2.5 L
4 x 2.5 L
6 x 1 L
Descripción
- For gas chromatography (GC)
- Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
- Non-volatile matter and water content are specified for each solvent
Especificaciones
Nombre del producto químico o material | Diethyl Ether |
Color | Colorless |
Fórmula lineal | (CH3CH2)2O |
Número UN | 1155 |
Beilstein | 1696894 |
Formula Weight (peso de la fórmula) | 74.12g/mol |
Grado | CHROMASOLV™ |
Densidad | 0.706g/mL at 25°C |
Cantidad | 4 x 2.5 L |
Límite de explosión | 0.365 |
Mostrar más |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'