missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hidrógeno-ortofosfato diamónico, certificado AR para análisis, Fisher Chemical
Click to view available options
Cantidad:
500 g
1 kg
Descripción
Di-Ammonium Hydrogen Orthophosphate, Certified AR for analysis
Especificaciones
Color | White |
pH | 7.9 to 8.3 |
Forma física | Crystalline Solid |
Cantidad | 1 kg |
Formula Weight (peso de la fórmula) | 132.06g/mol |
Envase | Plastic powder jar |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'