Learn More
Thermo Scientific Chemicals [Des-octanoyl]-Ghrelin, Human
For cellular and molecular biology applications
Marca: Thermo Scientific™ J65946.M^
Detalles adicionales : CAS : 313951-59-6 Peso : 0.00001kg
Descripción
Inhibits cell proliferation in breast and prostate cancer cell lines. Promotes adipogenesis in vivo.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Human | |
-30°C to -10°C | |
3244.66 | |
1 mg | |
Gly-Ser-Ser-Phe-Leu-Ser-Pro-Glu-His-Gln-Arg-Val-Gln-Gln-Arg-Lys-Glu-Ser-Lys-Lys-Pro-Pro-Ala-Lys-Leu-Gln-Pro-Arg |
HPLC | |
Solid | |
[Des-octanoyl]-Ghrelin, human | |
RUO - research use only |
RUO – Research Use Only