missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DBT Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 12451-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
The branched-chain alpha-keto acid dehydrogenase complex (BCKD) is an inner-mitochondrial enzyme complex involved in the breakdown of the branched-chain amino acids isoleucine, leucine, and valine. The BCKD complex is thought to be composed of a core of 24 transacylase (E2) subunits, and associated decarboxylase (E1), dehydrogenase (E3), and regulatory subunits. This gene encodes the transacylase (E2) subunit. Mutations in this gene result in maple syrup urine disease, type 2. Alternatively spliced transcript variants have been described, but their biological validity has not been determined.Especificaciones
DBT | |
Polyclonal | |
Unconjugated | |
DBT | |
DBT, E2, E2B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
13171, 1629, 29611 | |
-20°C | |
Liquid |
Western Blot, Immunofluorescence, Immunocytochemistry | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P11182, P53395 | |
Dbt | |
DBT Fusion Protein Ag3101 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |