missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DAP5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 17728-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
CDK5RAP2 is a regulator of CDK5 (cyclin-dependent kinase 5) activity. The protein is localized to the centrosome and Golgi complex and interacts with CDK5R1 and pericentrin (PCNT). CDK5RAP2 plays a role in centriole engagement and microtubule nucleation, and has been linked to primary microcephaly and Alzheimer′s disease. Alternative splicing results in multiple transcript variants.Especificaciones
DAP5 | |
Polyclonal | |
Unconjugated | |
EIF4G2 | |
AAG1, DAP 5, DAP5, Death associated protein 5, eIF 4 gamma 2, eIF 4G 2, eIF4G 2, EIF4G2, NAT1, p97 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
13690, 1982, 361628 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P78344, Q62448 | |
Eif4g2 | |
DAP5 Fusion Protein Ag12125 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |