Learn More
Thermo Scientific™ [D-Arg1, D-Phe5, D-Trp7,9, Leu11] Substance P
For cellular and molecular biology applications
Marca: Thermo Scientific™ J66988.M¬
Detalles adicionales : CAS : 96736-12-8 Peso : 0.00001kg
Descripción
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Especificaciones
HPLC | |
-30°C to -10°C | |
1516.85 | |
1 mg | |
D-Arg-Pro-Lys-Pro-D-Phe-Gln-D-Trp-Phe-D-Trp-Leu-Leu-NH2 |
Antagonist D | |
Solid | |
[D-Arg1, D-Phe5, D-Trp7,9, Leu11] Substance P | |
RUO - research use only |
For Research Use Only