Learn More
Cytokeratin 8 Ab-4 Mouse Monoclonal Antibody, Epredia™
Ensure accurate, reproducible results in immunohistochemistry and immunoprecipitation experiments with Epredia™ Cytokeratin 8 Ab-4, Mouse Monoclonal Antibody.
Marca: Epredia MS-997-B
Detalles adicionales : Peso : 0.00100kg
Beschreibung
Keratin 8 belongs to the type B (basic) subfamily of high molecular weight keratins and exists in combination with keratin 18. Keratin 8 is primarily found in the non-squamous epithelia and is present in majority of adenocarcinomas and ductal carcinomas. It is absent in squamous cell carcinomas. Hepatocellular carcinomas are defined by the use of antibodies that recognize only cytokeratin polypeptides 8 and 18.Host Species: Mouse
Clone: TS1
Isotype: IgG1/κ
Species Reactivity: Human. Does not react with rat. Others not known.
Epitope: aa 343-357
Immunogen: Keratin preparation from a human carcinoma
Molecular Weight: 52.5 kDa
Positive Control: MCF-7 or A431 cells; Skin; Colon, Lung or Breast CA
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Immunoprecipitation
Spezifikation
Cytokeratin 8 Ab-4 | |
Monoclonal | |
200μg/mL | |
Mouse | |
1 mL | |
Cytoskeletal Protein | |
Human | |
IgG2a κ |
Immunohistochemistry (Paraffin), Immunoprecipitation | |
TS1 | |
Biotin | |
Keratin preparation from a human carcinoma | |
IVD | |
Primary | |
Concentrated |