missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYC1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 10242-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This is the heme-containing component of the cytochrome b-c1 complex, which accepts electrons from Rieske protein and transfers electrons to cytochrome c in the mitochondrial respiratory chain.Especificaciones
CYC1 | |
Polyclonal | |
Unconjugated | |
CYC1 | |
Complex III subunit 4, Complex III subunit IV, CYC1, cytochrome c 1, UQCR4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1537, 66445 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P08574, Q9D0M3 | |
Cyc1 | |
CYC1 Fusion Protein Ag0291 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |