missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CSNK1A1L Recombinant Human Protein
This CSNK1A1L Recombinant Human Protein (full length) was expressed in insect cells.
Marca: Invitrogen™ A30494
Detalles adicionales : Peso : 0.10000kg
Descripción
- This CSNK1A1L recombinant human protein (full length) was expressed in insect cells
- CSNK1A1L serine/threonine kinase, or CK1-alpha, dynamically associates with the CBM complex upon T cell receptor engagement to participate in cytokine production
- In lymphocyte proliferation it has a contrasting role by subsequently promoting the phosphorylation and inactivation of CARMA1
- It governs antigen-receptor-induced NF-kappa-B activation and human lymphoma cell survival
- CSNK1A1L recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service
- Validated for use with the Z’-LYTE™ assay
- Available in multiple pack sizes for convenient use
Especificaciones
AAH28723 | |
10 µg | |
Full Length | |
GST Tag | |
Dry Ice | |
Full Length, Recombinant, Active | |
Kinase Assay |
65.4 kDa | |
CK1 Kinase Family | |
Yes | |
Storage Buffer: 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol. | |
Insect | |
Liquid | |
Human |