missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fosfato de creatina, sal disódica tetrahidrato, ≥ 98 %, Affymetrix/USB™
Click to view available options
Cantidad:
5 g
25 g
Envase:
Frasco de plástico
Descripción
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Especificaciones
Nombre del producto químico o material | Creatine Phosphate Disodium Salt Tetrahydrate |
Forma física | Polvo cristalino |
Cantidad | 5 g |
Formula Weight (peso de la fórmula) | 327.14 |
Porcentaje de pureza | ≥98% |
Envase | Frasco de plástico |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'