missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper(Ii) Sulfate Pentahydrate, ACS Reagent, Honeywell Fluka™
Click to view available options
Cantidad:
100 g
250 g
500 g
2.5 kg
Descripción
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Especificaciones
Nombre del producto químico o material | Copper(Ii) Sulfate Pentahydrate |
Cantidad | 500 g |
Fórmula lineal | CuSO4 · 5H2O |
Número UN | 3077 |
Nombre IUPAC | copper;sulfate;pentahydrate |
Formula Weight (peso de la fórmula) | 249.69g/mol |
Porcentaje de pureza | ≥98% |
Grado | ACS Reagent |
Presión de vapor | 7.3mmHg (25°C) |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'