missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper(Ii) Acetate Monohydrate, Honeywell™
Click to view available options
Cantidad:
500 g
1 kg
50 kg
6 x 1 kg
Descripción
- We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Especificaciones
Nombre del producto químico o material | Copper(Ii) Acetate Monohydrate |
Cantidad | 6 x 1 kg |
Fórmula lineal | Cu(CO2CH3)2 · H2O |
Número UN | UN1759 |
Nombre IUPAC | copper(2+) diacetate hydrate |
Formula Weight (peso de la fórmula) | 199.65 |
Porcentaje de pureza | ≥99% (iodometric) |
Densidad | Vapor Density: 6.8 (vs air) |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'