Learn More
Connexin 43 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 26980-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene is a member of the connexin gene family. The encoded protein is a component of gap junctions, which are composed of arrays of intercellular channels that provide a route for the diffusion of low molecular weight materials from cell to cell. The encoded protein is the major protein of gap junctions in the heart that are thought to have a crucial role in the synchronized contraction of the heart and in embryonic development. A related intronless pseudogene has been mapped to chromosome 5. Mutations in this gene have been associated with oculodentodigital dysplasia and heart malformations.Especificaciones
Connexin 43 | |
Polyclonal | |
Unconjugated | |
GJA1 | |
Connexin 43, CX43, DFNB38, Gap junction alpha 1 protein, GJA1, GJAL, ODDD | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14609, 2697 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P17302, P23242 | |
Gja1 | |
Connexin 43 Fusion Protein Ag25533 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |