missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement factor B Mouse anti-Human, Clone: 8D2B6, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66154-1-IG-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes complement factor B, a component of the alternative pathway of complement activation. Factor B circulates in the blood as a single chain polypeptide. Upon activation of the alternative pathway, it is cleaved by complement factor D yielding the noncatalytic chain Ba and the catalytic subunit Bb. The active subunit Bb is a serine protease which associates with C3b to form the alternative pathway C3 convertase. Bb is involved in the proliferation of preactivated B lymphocytes, while Ba inhibits their proliferation. This gene localizes to the major histocompatibility complex (MHC) class III region on chromosome 6. This cluster includes several genes involved in regulation of the immune reaction. Polymorphisms in this gene are associated with a reduced risk of age-related macular degeneration. The polyadenylation site of this gene is 421 bp from the 5′ end of the gene for complement component 2.Especificaciones
Complement factor B | |
Monoclonal | |
0.6 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P00751 | |
CFB | |
Complement factor B Fusion Protein Ag19107 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgM |
Western Blot, Immunohistochemistry (Paraffin) | |
8D2B6 | |
Unconjugated | |
CFB | |
BF, BFD, C3/C5 convertase, CFAB, CFB, Complement factor B, FB, FBI12, GBG, Glycine rich beta glycoprotein, H2 Bf, PBF2, Properdin factor B | |
Mouse | |
Antigen Affinity Chromatography | |
RUO | |
629 | |
-20°C | |
Liquid |