missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLPP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 15698-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
The protein encoded by this gene belongs to the peptidase family S14 and hydrolyzes proteins into small peptides in the presence of ATP and magnesium. The protein is transported into mitochondrial matrix and is associated with the inner mitochondrial membrane.Especificaciones
CLPP | |
Polyclonal | |
Unconjugated | |
CLPP | |
CLPP, Endopeptidase Clp | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
301117, 53895, 8192 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunoprecipitation, Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin) | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O88696, Q16740 | |
Clpp | |
CLPP Fusion Protein Ag8325 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |