missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human CHUK (IKK Alpha), His Tag Recombinant Protein
Human CHUK (IKK Alpha), His Tag Recombinant Protein
Marca: Invitrogen™ PV4310
Detalles adicionales : Peso : 0.05250kg
Descripción
CHUK (IKK alpha) is a serine⁄threonine kinase involved in signal transduction and cellular communication. It belongs to the cytokine activated protein complex that is an inhibitor of the NF-kappa B complex.Especificaciones
NP_001269 | |
114294, 1147 | |
10 μg | |
IKK Kinase Family | |
Recombinant | |
Storage Buffer: 50 mM Tris-HCl pH 7.5, 150 mM NaCl, 0.5 mM EDTA, 0.02% Triton™ X-100, 2 mM DTT, and 50% Glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Ser⁄Thr Kinases: Other | |
LanthaScreen™ Antibody, LanthaScreen™ Substrate, Adapta™ | |
conserved helix-loop-helix ubiquitous kinase | |
Kinases & Inhibitors | |
Liquid | |
Signal Transduction |
Kinase Assay | |
88.7 kDa | |
CHUK (IKK α) | |
Human | |
His Tag | |
Biochemical terbium phospho-specific activity, Biochemical binding/kinetic | |
Dry Ice | |
CHUK | |
Baculovirus | |
Recombinant, Full Length | |
Serine/Threonine Kinases |