missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHSY1 Rabbit anti-Human, Mouse, Rat, Zebrafish, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 14420-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
CHSY1 synthesizes chondroitin sulfate, a glycosaminoglycan expressed on the surface of most cells and in extracellular matrices. Glycosaminoglycan chains are covalently linked to a wide range of core protein families and regulate many biologic processes, including cell proliferation and recognition, extracellular matrix deposition, and morphogenesis.Especificaciones
CHSY1 | |
Polyclonal | |
Unconjugated | |
CHSY1 | |
chondroitin sulfate synthase 1, Chondroitin synthase 1, CHSY, ChSy 1, CHSY1, CSS1, KIAA0990 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
22856, 269941, 292999, 324407 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.39 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6ZQ11, Q86X52 | |
chsy1 | |
CHSY1 Fusion Protein Ag5766 | |
150 μL | |
Primary | |
Human, Mouse, Rat, Zebrafish | |
Antibody | |
IgG |