missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CES7 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 19520-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
The antibody can recognize the isoform1 and isoform2.
CES7 is involved in the detoxification of xenobiotics and in the activation of ester and amide prodrugs.Especificaciones
CES7 | |
Polyclonal | |
Unconjugated | |
CES5A | |
Carboxylesterase 7, CAUXIN, CES4C1, CES5, CES7 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
221223 | |
-20°C | |
Liquid |
Immunocytochemistry, Western Blot, Immunofluorescence | |
0.46 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6NT32 | |
CES5A | |
Peptide | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |